2-Chloro-N-(4-methoxybenzyl)acetamide

Supplier: Matrix Scientific

029634-500MG
101828-550EA 143.08 USD
101828-550
2-Chloro-N-(4-methoxybenzyl)acetamide
2-Chloro-N-(4-methoxybenzyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₂
MDL Number: MFCD02668858
CAS Number: 81494-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR