4-[(Chloroacetyl)amino]-N-isopropylbenzamide

Supplier: Matrix Scientific

029632-500MG
101828-546EA 290.77 USD
101828-546
4-[(Chloroacetyl)amino]-N-isopropylbenzamide
4-[(Chloroacetyl)amino]-N-isopropylbenzamide

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O₂
MDL Number: MFCD08593593
CAS Number: 908518-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR