2-Chloro-N-(3,4-dichlorobenzyl)acetamide

Supplier: Matrix Scientific

029630-500MG
101828-542EA 221.54 USD
101828-542
2-Chloro-N-(3,4-dichlorobenzyl)acetamide
2-Chloro-N-(3,4-dichlorobenzyl)acetamide

Hazard: Irritant

Formula: C₉H₈Cl₃NO
MDL Number: MFCD03362899
CAS Number: 56978-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR