2-[(Chloroacetyl)amino]-4,5-dimethylthiophene-3-carboxamide

Supplier: Matrix Scientific

029628-500MG
101828-538EA 217.35 USD
101828-538
2-[(Chloroacetyl)amino]-4,5-dimethylthiophene-3-carboxamide
2-[(Chloroacetyl)amino]-4,5-dimethylthiophene-3-carboxamide

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂S
MDL Number: MFCD01355935
CAS Number: 551903-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR