Ethyl-2-[(chloroacetyl)amino]-5-[(dimethylamino)carbonyl]-4-methylthiophene-3-carboxylate
Supplier: Matrix Scientific
029626-500MG
101828-534EA
217.35
USD
101828-534
Ethyl-2-[(chloroacetyl)amino]-5-[(dimethylamino)carbonyl]-4-methylthiophene-3-carboxylate
Ethyl-2-[(chloroacetyl)amino]-5-[(dimethylamino)carbonyl]-4-methylthiophene-3-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₇ClN₂O₄S |
MDL Number:
MFCD03396357 CAS Number: 546116-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...