Ethyl-2-[(chloroacetyl)amino]-5-phenylthiophene-3-carboxylate

Supplier: Matrix Scientific

029625-500MG
101828-532EA 217.35 USD
101828-532
Ethyl-2-[(chloroacetyl)amino]-5-phenylthiophene-3-carboxylate
Ethyl-2-[(chloroacetyl)amino]-5-phenylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₄ClNO₃S
MDL Number: MFCD02350907
CAS Number: 379256-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR