4-[(Chloroacetyl)amino]-N-methylbenzamide

Supplier: Matrix Scientific

029623-500MG
101828-528EA 146.15 USD
101828-528
4-[(Chloroacetyl)amino]-N-methylbenzamide
4-[(Chloroacetyl)amino]-N-methylbenzamide

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂O₂
MDL Number: MFCD07348598
CAS Number: 871217-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR