2-Chloro-N-(2,5-difluorophenyl)acetamide

Supplier: Matrix Scientific

Synonyms: N-Chloroacetyl-2,5-difluoroaniline

029622-500MG
101828-526EA 146.15 USD
101828-526
2-Chloro-N-(2,5-difluorophenyl)acetamide
2-Chloro-N-(2,5-difluorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₆ClF₂NO
MW: 205.59 g/mol
MDL Number: MFCD00157387
CAS Number: 196938-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR