N-2,1,3-Benzoxadiazol-4-yl-2-chloroacetamide

Supplier: Matrix Scientific

029621-500MG
101828-524EA 217.35 USD
101828-524
N-2,1,3-Benzoxadiazol-4-yl-2-chloroacetamide
N-2,1,3-Benzoxadiazol-4-yl-2-chloroacetamide

Hazard: Irritant

Formula: C₈H₆ClN₃O₂
MDL Number: MFCD08593591
CAS Number: 908518-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR