4-[(Chloroacetyl)amino]-N-cyclopentylbenzamide

Supplier: Matrix Scientific

029620-500MG
101828-522EA 217.35 USD
101828-522
4-[(Chloroacetyl)amino]-N-cyclopentylbenzamide
4-[(Chloroacetyl)amino]-N-cyclopentylbenzamide

Hazard: Irritant

Formula: C₁₄H₁₇ClN₂O₂
MDL Number: MFCD08593590
CAS Number: 908518-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR