Ethyl-2-[(chloroacetyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

029619-500MG
101828-520EA 338.46 USD
101828-520
Ethyl-2-[(chloroacetyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate
Ethyl-2-[(chloroacetyl)amino]-4-ethyl-5-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₃S
MDL Number: MFCD01336543
CAS Number: 565191-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR