5-[(Chloroacetyl)amino]-4-cyano-N,N-diethyl-3-methylthiophene-2-carboxamide

Supplier: Matrix Scientific

029615-500MG
101828-512EA 217.35 USD
101828-512
5-[(Chloroacetyl)amino]-4-cyano-N,N-diethyl-3-methylthiophene-2-carboxamide
5-[(Chloroacetyl)amino]-4-cyano-N,N-diethyl-3-methylthiophene-2-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₆ClN₃O₂S
MDL Number: MFCD03363526
CAS Number: 540793-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR