N-(3-Bromophenyl)-2-chloroacetamide

Supplier: Matrix Scientific

029609-500MG
101828-500EA 146.15 USD
101828-500
N-(3-Bromophenyl)-2-chloroacetamide
N-(3-Bromophenyl)-2-chloroacetamide

Hazard: Irritant

Formula: C₈H₇BrClNO
Melting Pt: 113…114 °C
MDL Number: MFCD00028175
CAS Number: 2564-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR