2-Chloro-N-(2-methoxy-5-methylphenyl)acetamide

Supplier: Matrix Scientific

029606-500MG
101828-494EA 146.15 USD
101828-494
2-Chloro-N-(2-methoxy-5-methylphenyl)acetamide
2-Chloro-N-(2-methoxy-5-methylphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₂
MDL Number: MFCD00095382
CAS Number: 85817-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR