2-Chloro-N-(2-fluorobenzyl)acetamide

Supplier: Matrix Scientific

029603-500MG
101828-488EA 193.85 USD
101828-488
2-Chloro-N-(2-fluorobenzyl)acetamide
2-Chloro-N-(2-fluorobenzyl)acetamide

Hazard: Irritant

Formula: C₉H₉ClFNO
MDL Number: MFCD08076774
CAS Number: 895367-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR