2-Chloro-N-(3,4,5-trimethoxyphenyl)acetamide
Supplier: Matrix Scientific
029602-500MG
101828-486EA
146.15
USD
101828-486
2-Chloro-N-(3,4,5-trimethoxyphenyl)acetamide
2-Chloro-N-(3,4,5-trimethoxyphenyl)acetamide
Hazard: Irritant
Formula:
C₁₁H₁₄ClNO₄ |
MDL Number:
MFCD00524910 CAS Number: 39901-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...