N-(4-Bromo-3-methylphenyl)-2-chloroacetamide

Supplier: Matrix Scientific

029599-500MG
101828-480EA 146.15 USD
101828-480
N-(4-Bromo-3-methylphenyl)-2-chloroacetamide
N-(4-Bromo-3-methylphenyl)-2-chloroacetamide

Hazard: Irritant

Formula: C₉H₉BrClNO
MDL Number: MFCD03147342
CAS Number: 98028-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR