N-(2-Bromo-4-methylphenyl)-2-chloroacetamide

Supplier: Matrix Scientific

029598-500MG
101828-478EA 146.15 USD
101828-478
N-(2-Bromo-4-methylphenyl)-2-chloroacetamide
N-(2-Bromo-4-methylphenyl)-2-chloroacetamide

Hazard: Irritant

Formula: C₉H₉BrClNO
MDL Number: MFCD00709711
CAS Number: 90560-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR