4-[(Chloroacetyl)amino]benzamide

Supplier: Matrix Scientific

029593-500MG
101828-468EA 230.77 USD
101828-468
4-[(Chloroacetyl)amino]benzamide
4-[(Chloroacetyl)amino]benzamide

Hazard: Irritant

Formula: C₉H₉ClN₂O₂
MDL Number: MFCD00566630
CAS Number: 85126-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR