2-Chloro-N-(3-chloro-4-methylphenyl)acetamide

Supplier: Matrix Scientific

029591-500MG
101828-464EA 109.25 USD
101828-464
2-Chloro-N-(3-chloro-4-methylphenyl)acetamide
2-Chloro-N-(3-chloro-4-methylphenyl)acetamide

Hazard: Irritant

Formula: C₉H₉Cl₂NO
MDL Number: MFCD00032562
CAS Number: 99585-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR