2-Chloro-N-(5-chloro-2,4-dimethoxyphenyl)acetamide
Supplier: Matrix Scientific
029587-500MG
101828-456EA
146.15
USD
101828-456
2-Chloro-N-(5-chloro-2,4-dimethoxyphenyl)acetamide
2-Chloro-N-(5-chloro-2,4-dimethoxyphenyl)acetamide
Hazard: Irritant
Formula:
C₁₀H₁₁Cl₂NO₃ |
MDL Number:
MFCD00157381 CAS Number: 23543-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...