Methyl-2-[(chloroacetyl)amino]-4-methyl-1,3-thiazole-5-carboxylate

Supplier: Matrix Scientific

029581-500MG
101828-444EA 290.77 USD
101828-444
Methyl-2-[(chloroacetyl)amino]-4-methyl-1,3-thiazole-5-carboxylate
Methyl-2-[(chloroacetyl)amino]-4-methyl-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₈H₉ClN₂O₃S
MDL Number: MFCD03392196
CAS Number: 6125-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR