2-Chloro-N-(4-chloro-2-fluorophenyl)acetamide

Supplier: Matrix Scientific

029579-500MG
101828-440EA 109.25 USD
101828-440
2-Chloro-N-(4-chloro-2-fluorophenyl)acetamide
2-Chloro-N-(4-chloro-2-fluorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₆Cl₂FNO
MDL Number: MFCD02725316
CAS Number: 380345-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR