2-Chloro-N-(2,6-dichlorophenyl)propanamide

Supplier: Matrix Scientific

029576-500MG
101828-434EA 146.15 USD
101828-434
2-Chloro-N-(2,6-dichlorophenyl)propanamide
2-Chloro-N-(2,6-dichlorophenyl)propanamide

Hazard: Irritant

Formula: C₉H₈Cl₃NO
MDL Number: MFCD08444157
CAS Number: 42276-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR