Dimethyl-2-[(2-chloropropanoyl)amino]terephthalate

Supplier: Matrix Scientific

029568-500MG
101828-418EA 146.15 USD
101828-418
Dimethyl-2-[(2-chloropropanoyl)amino]terephthalate
Dimethyl-2-[(2-chloropropanoyl)amino]terephthalate

Hazard: Irritant

Formula: C₁₃H₁₄ClNO₅
MDL Number: MFCD09971921
CAS Number: 956576-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR