N-(3-Acetylphenyl)-2-chloropropanamide

Supplier: Matrix Scientific

029567-500MG
101828-416EA 109.25 USD
101828-416
N-(3-Acetylphenyl)-2-chloropropanamide
N-(3-Acetylphenyl)-2-chloropropanamide

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₂
MDL Number: MFCD03147407
CAS Number: 379255-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR