2-Chloro-N-(4-ethoxyphenyl)propanamide

Supplier: Matrix Scientific

029565-500MG
101828-412EA 146.15 USD
101828-412
2-Chloro-N-(4-ethoxyphenyl)propanamide
2-Chloro-N-(4-ethoxyphenyl)propanamide

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₂
MDL Number: MFCD08444156
CAS Number: 868771-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR