2-Chloro-N-(2,4-dimethoxyphenyl)propanamide

Supplier: Matrix Scientific

029559-500MG
101828-400EA 109.25 USD
101828-400
2-Chloro-N-(2,4-dimethoxyphenyl)propanamide
2-Chloro-N-(2,4-dimethoxyphenyl)propanamide

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₃
MDL Number: MFCD09932050
CAS Number: 956576-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR