2-Chloro-N-(4-methoxyphenyl)propanamide

Supplier: Matrix Scientific

029558-500MG
101828-398EA 146.15 USD
101828-398
2-Chloro-N-(4-methoxyphenyl)propanamide
2-Chloro-N-(4-methoxyphenyl)propanamide

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₂
MDL Number: MFCD05666377
CAS Number: 21262-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR