2-Chloro-N-(3-chloro-2-methylphenyl)propanamide

Supplier: Matrix Scientific

029557-500MG
101828-396EA 146.15 USD
101828-396
2-Chloro-N-(3-chloro-2-methylphenyl)propanamide
2-Chloro-N-(3-chloro-2-methylphenyl)propanamide

Hazard: Irritant

Formula: C₁₀H₁₁Cl₂NO
MDL Number: MFCD09397813
CAS Number: 930395-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR