2-Chloro-N-(2,5-dimethylphenyl)propanamide

Supplier: Matrix Scientific

029553-500MG
101828-388EA 146.15 USD
101828-388
2-Chloro-N-(2,5-dimethylphenyl)propanamide
2-Chloro-N-(2,5-dimethylphenyl)propanamide

Hazard: Irritant

Formula: C₁₁H₁₄ClNO
MDL Number: MFCD09413165
CAS Number: 400755-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR