2-Chloro-N-(3,4-dichlorophenyl)propanamide
Supplier: Matrix Scientific
029547-500MG
101828-376EA
146.15
USD
101828-376
2-Chloro-N-(3,4-dichlorophenyl)propanamide
2-Chloro-N-(3,4-dichlorophenyl)propanamide
Hazard: Irritant
Formula:
C₉H₈Cl₃NO |
MDL Number:
MFCD09971920 CAS Number: 25251-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...