2-Chloro-N-(2,5-dichlorophenyl)propanamide

Supplier: Matrix Scientific

029545-500MG
101828-372EA 146.15 USD
101828-372
2-Chloro-N-(2,5-dichlorophenyl)propanamide
2-Chloro-N-(2,5-dichlorophenyl)propanamide

Hazard: Irritant

Formula: C₉H₈Cl₃NO
MDL Number: MFCD03988550
CAS Number: 554439-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR