N-(4-Bromophenyl)-2-chloropropanamide

Supplier: Matrix Scientific

029539-500MG
101828-360EA 109.25 USD
101828-360
N-(4-Bromophenyl)-2-chloropropanamide
N-(4-Bromophenyl)-2-chloropropanamide

Hazard: Irritant

Formula: C₉H₉BrClNO
MDL Number: MFCD03984459
CAS Number: 21262-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR