2-Chloro-N-(4-chlorophenyl)propanamide
Supplier: Matrix Scientific
029538-500MG
101828-358EA
146.15
USD
101828-358
2-Chloro-N-(4-chlorophenyl)propanamide
2-Chloro-N-(4-chlorophenyl)propanamide
Hazard: Irritant
Formula:
C₉H₉Cl₂NO |
MDL Number:
MFCD04621521 CAS Number: 21262-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...