7-(Chloromethyl)-3-phenyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one
Supplier: Matrix Scientific
029536-500MG
101828-354EA
307.69
USD
101828-354
7-(Chloromethyl)-3-phenyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one
7-(Chloromethyl)-3-phenyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one
Hazard: Irritant
Formula:
C₁₃H₉ClN₂OS MW: 276.75 g/mol |
MDL Number:
MFCD02059663 CAS Number: 85811-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...