4-(Chloromethyl)-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole

Supplier: Matrix Scientific

029532-500MG
101828-346EA 290.77 USD
101828-346
4-(Chloromethyl)-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole
4-(Chloromethyl)-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole

Hazard: Irritant

Formula: C₁₁H₇ClF₃NS
MDL Number: MFCD07612585
CAS Number: 886629-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR