2-(4-Bromophenyl)-4-(chloromethyl)-1,3-thiazole

Supplier: Matrix Scientific

029531-500MG
101828-344EA 217.35 USD
101828-344
2-(4-Bromophenyl)-4-(chloromethyl)-1,3-thiazole
2-(4-Bromophenyl)-4-(chloromethyl)-1,3-thiazole

Hazard: Irritant

Formula: C₁₀H₇BrClNS
MDL Number: MFCD09040953
CAS Number: 35199-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR