4-(Chloromethyl)-2-(4-fluorophenyl)-1,3-thiazole

Supplier: Matrix Scientific

029530-500MG
101828-342EA 290.77 USD
101828-342
4-(Chloromethyl)-2-(4-fluorophenyl)-1,3-thiazole
4-(Chloromethyl)-2-(4-fluorophenyl)-1,3-thiazole

Hazard: Irritant

Formula: C₁₀H₇ClFNS
MDL Number: MFCD05273594
CAS Number: 113264-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR