4-(Chloromethyl)-2-(4-chlorophenyl)-1,3-thiazole

Supplier: Matrix Scientific

029529-500MG
101828-340EA 290.77 USD
101828-340
4-(Chloromethyl)-2-(4-chlorophenyl)-1,3-thiazole
4-(Chloromethyl)-2-(4-chlorophenyl)-1,3-thiazole

Hazard: Irritant

Formula: C₁₀H₇Cl₂NS
MDL Number: MFCD00047057
CAS Number: 17969-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR