2-Chloro-N-(3-cyano-6-ethyl-4,5,6,7-tetrahydro-1-benzothien-2-yl)acetamide

Supplier: Matrix Scientific

029523-500MG
101828-328EA 290.77 USD
101828-328
2-Chloro-N-(3-cyano-6-ethyl-4,5,6,7-tetrahydro-1-benzothien-2-yl)acetamide
2-Chloro-N-(3-cyano-6-ethyl-4,5,6,7-tetrahydro-1-benzothien-2-yl)acetamide

Hazard: Irritant

Formula: C₁₃H₁₅ClN₂OS
MDL Number: MFCD01354872
CAS Number: 551899-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR