2-Chloro-N-(2-chloropyridin-3-yl)acetamide
Supplier: Matrix Scientific
029521-500MG
101828-324EA
290.77
USD
101828-324
2-Chloro-N-(2-chloropyridin-3-yl)acetamide
2-Chloro-N-(2-chloropyridin-3-yl)acetamide
Hazard: Irritant
Formula:
C₇H₆Cl₂N₂O |
MDL Number:
MFCD00104119 CAS Number: 379726-55-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...