Methyl-2-[(chloroacetyl)amino]thiophene-3-carboxylate

Supplier: Matrix Scientific

029520-500MG
101828-322EA 233.85 USD
101828-322
Methyl-2-[(chloroacetyl)amino]thiophene-3-carboxylate
Methyl-2-[(chloroacetyl)amino]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₈H₈ClNO₃S
MDL Number: MFCD00661989
CAS Number: 590355-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR