2-Chloro-N-(3-cyanothien-2-yl)acetamide

Supplier: Matrix Scientific

029519-500MG
101828-320EA 290.77 USD
101828-320
2-Chloro-N-(3-cyanothien-2-yl)acetamide
2-Chloro-N-(3-cyanothien-2-yl)acetamide

Hazard: Irritant

Formula: C₇H₅ClN₂OS
MDL Number: MFCD00112765
CAS Number: 55654-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR