2-(1,3-Benzodioxol-5-yl)-4-(chloromethyl)-1,3-thiazole

Supplier: Matrix Scientific

029518-500MG
101828-318EA 290.77 USD
101828-318
2-(1,3-Benzodioxol-5-yl)-4-(chloromethyl)-1,3-thiazole
2-(1,3-Benzodioxol-5-yl)-4-(chloromethyl)-1,3-thiazole

Hazard: Irritant

Formula: C₁₁H₈ClNO₂S
MDL Number: MFCD04054556
CAS Number: 55315-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR