2-Chloro-N-[4-(6-methyl-1,3-benzothiazol-2-yl)phenyl]acetamide

Supplier: Matrix Scientific

029517-500MG
101828-316EA 307.69 USD
101828-316
2-Chloro-N-[4-(6-methyl-1,3-benzothiazol-2-yl)phenyl]acetamide
2-Chloro-N-[4-(6-methyl-1,3-benzothiazol-2-yl)phenyl]acetamide

Hazard: Irritant

Formula: C₁₆H₁₃ClN₂OS
MDL Number: MFCD00709263
CAS Number: 87992-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR