2-Chloro-N-phenethylacetamide

Supplier: Matrix Scientific

029515-500MG
101828-312EA 109.25 USD
101828-312
2-Chloro-N-phenethylacetamide
2-Chloro-N-phenethylacetamide

Hazard: Irritant

Formula: C₁₀H₁₂ClNO
MW: 197.67 g/mol
MDL Number: MFCD00245084
CAS Number: 13156-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR