2-Chloro-N-cycloheptylacetamide

Supplier: Matrix Scientific

029514-500MG
101828-310EA 336.92 USD
101828-310
2-Chloro-N-cycloheptylacetamide
2-Chloro-N-cycloheptylacetamide

Hazard: Irritant

Formula: C₉H₁₆ClNO
MDL Number: MFCD01343566
CAS Number: 23511-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR