2-Chloro-N-(2,4,5-trichlorophenyl)acetamide

Supplier: Matrix Scientific

029511-500MG
101828-304EA 146.15 USD
101828-304
2-Chloro-N-(2,4,5-trichlorophenyl)acetamide
2-Chloro-N-(2,4,5-trichlorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₅Cl₄NO
MDL Number: MFCD00018889
CAS Number: 23595-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR