2-Chloro-N-(2,6-dichlorophenyl)acetamide

Supplier: Matrix Scientific

029510-500MG
101828-302EA 146.15 USD
101828-302
2-Chloro-N-(2,6-dichlorophenyl)acetamide
2-Chloro-N-(2,6-dichlorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₆Cl₃NO
MDL Number: MFCD00044773
CAS Number: 3644-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR